6,7,8,9-Tetrahydro-5H-benzocyclohepten-5-one ≥97.0% (by GC)
Fornitore: TCI
T1347-25G
T1347-5G
TCIAT1347-25GEA
274
EUR
TCIAT1347-25G
TCIAT1347-5G
6,7,8,9-Tetrahydro-5H-benzocyclohepten-5-one ≥97.0% (by GC)
6,7,8,9-Tetrahydro-5H-benzocyclohepten-5-one
Formula:
C₁₁H₁₂O Punto di ebollizione: 270°C |
MDL Number:
MFCD00004144 CAS Number: 826-73-3 EINECS: 212-558-4 |
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |