3-Trifluoromethyl-4'-methoxydiphenylamine
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
N-(4-Methoxyphenyl)-3-(trifluoromethyl)aniline, N-(3-Trifluoromethylphenyl)-p-anisidine
T1326-1G
T1326-5G
TCIAT1326-1GEA
94
EUR
TCIAT1326-1G
TCIAT1326-5G
3-Trifluoromethyl-4'-methoxydiphenylamine
3-Trifluoromethyl-4'-methoxydiphenylamine
Formula:
C₁₄H₁₂F₃NO Peso molecolare: 267,25 g/mol |
MDL Number:
MFCD00143198 CAS Number: 1494-26-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 56.0 to 61.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...