Stampa…

Triethanolamine borate ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,2',2''-Nitrilotriethyl Borate

T1291-25G T1291-500G T1291-100G
TCIAT1291-25GEA 34 EUR
TCIAT1291-25G TCIAT1291-500G TCIAT1291-100G
Triethanolamine borate ≥95.0% (by titrimetric analysis)
Triethanolamine borate
Formula: C₆H₁₂BNO₃
Peso molecolare: 156,98 g/mol
MDL Number: MFCD00003272
CAS Number: 283-56-7
EINECS: 206-003-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79fcb09037d38521b129d2c064dae5e0035b4c29&vid=79fcb09037d38521b129d2c064dae5e0035b4c29&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 95.0 %
Melting point 235.0 to 239.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR