Tetrabutylammonium triiodide ≥96.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Tetrabutylammonium Triiodide
T1271-25G
T1271-5G
TCIAT1271-25GEA
113
EUR
TCIAT1271-25G
TCIAT1271-5G
Tetrabutylammonium triiodide ≥96.0% (by titrimetric analysis)
Tetrabutylammonium triiodide
Formula:
C₁₆H₃₆I₃N Peso molecolare: 623,18 g/mol |
MDL Number:
MFCD00059123 CAS Number: 13311-45-0 |
Risultati del test di specificazione
Appearance | Gray to Dark purple to Black powder to crystal |
Purity (Iodometric Titration) | min. 96.0 % |
Melting point | 69.0 to 73.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...