Stampa…

1,1,1-Tris(4-hydroxyphenyl)ethane ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4,4',4''-(Ethane-1,1,1-triyl)triphenol

T1254-25G T1254-500G T1254-100G
TCIAT1254-25GEA 35 EUR
TCIAT1254-25G TCIAT1254-500G TCIAT1254-100G
1,1,1-Tris(4-hydroxyphenyl)ethane ≥98.0% (by GC)
1,1,1-Tris(4-hydroxyphenyl)ethane
Formula: C₂₀H₁₈O₃
Peso molecolare: 306,36 g/mol
Punto di fusione: 248°C
MDL Number: MFCD00012180
CAS Number: 27955-94-8
EINECS: 405-800-7
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caa433415b51ca61559c999839f735b8fbb2fae7&vid=caa433415b51ca61559c999839f735b8fbb2fae7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR