Stampa…

Trimethylsilylmethanol ≥97.0% (by GC)

Fornitore: TCI
Attenzione

T1250-25ML T1250-5ML
TCIAT1250-25MLEA 229 EUR
TCIAT1250-25ML TCIAT1250-5ML
Trimethylsilylmethanol ≥97.0% (by GC)
Trimethylsilylmethanol
Formula: C₄H₁₂OSi
Punto di ebollizione: 121°C
Flash Pt: 56°C
MDL Number: MFCD00004596
CAS Number: 3219-63-4
EINECS: 221-746-5
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4d6ba32b04604f2d46e295833c04efe37940149&vid=d4d6ba32b04604f2d46e295833c04efe37940149&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR