Tetrapropoxymethane ≥96.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Tetrapropyl Orthocarbonate
T1209-1ML
T1209-5ML
TCIAT1209-1MLEA
134
EUR
TCIAT1209-1ML
TCIAT1209-5ML
Tetrapropoxymethane ≥96.0% (by GC)
Tetrapropoxymethane
Formula:
C₁₃H₂₈O₄ Peso molecolare: 248,36 g/mol Punto di ebollizione: 224°C |
MDL Number:
MFCD00060101 CAS Number: 597-72-8 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...