Stampa…

L-Tyrosine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: L-Methyltyrosinate hydrochloride, H-Tyr-OMe.HCl, H-Tyr-OMe·HCl

T1108-25G T1108-5G
TCIAT1108-25GEA 45 EUR
TCIAT1108-25G TCIAT1108-5G
L-Tyrosine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
L-Tyrosine methyl ester hydrochloride
Formula: C₁₀H₁₃NO₃
Peso molecolare: 231,68 g/mol
MDL Number: MFCD00012607
CAS Number: 3417-91-2
EINECS: 222-313-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +12.0 to +13.0 deg (C=2, MeOH)
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR