Stampa…

Tris(dimethylamino)methane ≥97.0% (by titrimetric analysis) stabilizzato

Fornitore: TCI EUROPE
Pericolo


Stabilization: Stabilised with potassium hydroxide

T1057-25ML T1057-5ML
TCIAT1057-25MLEA 718 EUR
TCIAT1057-25ML TCIAT1057-5ML
Tris(dimethylamino)methane ≥97.0% (by titrimetric analysis) stabilizzato
Tris(dimethylamino)methane
Formula: C₇H₁₉N₃
Peso molecolare: 145,25 g/mol
Flash Pt: –15°C
MDL Number: MFCD00008322
CAS Number: 5762-56-1
EINECS: 227-284-0
UN: 2733
ADR: 3,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR