Trimethyl orthovalerate ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1,1,1-Trimethoxypentane, Orthovaleric Acid Trimethyl Ester
T0981-25ML
T0981-250ML
TCIAT0981-25MLEA
70
EUR
TCIAT0981-25ML
TCIAT0981-250ML
Trimethyl orthovalerate ≥97.0% (by GC)
Trimethyl orthovalerate
Formula:
C₈H₁₈O₃ Peso molecolare: 162,23 g/mol Punto di ebollizione: 164°C Flash Pt: 55°C |
MDL Number:
MFCD00008481 CAS Number: 13820-09-2 EINECS: 237-496-5 UN: 3272 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Specific gravity (20/20) | 0.9240 to 0.9260 |
Refractive index n20/D | 1.4090 to 1.4110 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...