Stampa…

Tetrahydrofuran-2,3,4,5-tetracarboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

T0975-25G T0975-500G
TCIAT0975-25GEA 112 EUR
TCIAT0975-25G TCIAT0975-500G
Tetrahydrofuran-2,3,4,5-tetracarboxylic acid ≥98.0% (by GC, titration analysis)
Tetrahydrofuran-2,3,4,5-tetracarboxylic acid
Formula: C₈H₈O₉
MDL Number: MFCD00005364
CAS Number: 26106-63-8
EINECS: 247-461-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR