Stampa…

1,2,4,5-Tetramethylimidazole ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,2,4,5-Tetramethyl-1H-imidazole

T0971-25G T0971-100G
TCIAT0971-25GEA 247 EUR
TCIAT0971-25G TCIAT0971-100G
1,2,4,5-Tetramethylimidazole ≥98.0% (by GC, titration analysis)
1,2,4,5-Tetramethylimidazole
Formula: C₇H₁₂N₂
Peso molecolare: 124,19 g/mol
MDL Number: MFCD00059162
CAS Number: 1739-83-9
EINECS: 217-100-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7a2945c65ca7475a8494a0b2e136e699c562425&vid=c7a2945c65ca7475a8494a0b2e136e699c562425&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR