Stampa…

Ethyl tetrahydrofuran-2-acetate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Tetrahydrofuran-2-acetic Acid Ethyl Ester

T0957-25ML T0957-5ML
TCIAT0957-25MLEA 518 EUR
TCIAT0957-25ML TCIAT0957-5ML
Ethyl tetrahydrofuran-2-acetate ≥98.0% (by GC)
Ethyl tetrahydrofuran-2-acetate
Formula: C₈H₁₄O₃
Peso molecolare: 158,20 g/mol
Punto di ebollizione: 98°C
MDL Number: MFCD00059735
CAS Number: 2434-02-8

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR