Stampa…

Trimethylolpropane trimethacrylate ≥90.0% (by GC) stabilizzato

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Ethyl-2-hydroxymethyl-1,3-propanediol Trimethacrylate, TMPTMA, 2-Ethyl-2-((methacryloyloxy)methyl)propane-1,3-diyl bis(2-methylacrylate)

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

T0912-25ML T0912-500ML
TCIAT0912-25MLEA 17 EUR
TCIAT0912-25ML TCIAT0912-500ML
Trimethylolpropane trimethacrylate ≥90.0% (by GC) stabilizzato
Trimethylolpropane trimethacrylate
Formula: C₁₈H₂₆O₆
Peso molecolare: 338,40 g/mol
Punto di fusione: –14°C
Flash Pt: 182°C
MDL Number: MFCD00008588
CAS Number: 3290-92-4
EINECS: 221-950-4

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR