Stampa…

Tamarind gum (from tamarind seed, polysaccharide) Polysaccharide

Fornitore: TCI EUROPE

T0909-500G T0909-25G
TCIAT0909-500GEA 95 EUR
TCIAT0909-500G TCIAT0909-25G
Tamarind gum (from tamarind seed, polysaccharide) Polysaccharide
Tamarind gum
MDL Number: MFCD00148561
CAS Number: 39386-78-2
EINECS: 254-442-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystaline
Arsenic max. 2.0ppm (as As2O3)
Heavy metals max. 20ppm (as Pb)
Protein max. 3.0%
Drying loss max. 7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR