Heptyl-p-toluenesulphonate ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₄H₂₂O₃S Punto di ebollizione: 201°C |
MDL Number:
MFCD00059442 CAS Number: 24767-82-6 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...