Stampa…
2-(4-Thiazolyl)benzimidazole ≥98.0% (by GC, titration analysis)
Fornitore: TCI
Pericolo
Synonyms: Thiabendazole
T0830-25G T0830-250G
TCIAT0830-25GEA 35 EUR
TCIAT0830-25G TCIAT0830-250G
2-(4-Thiazolyl)benzimidazole ≥98.0% (by GC, titration analysis)
2-(4-Thiazolyl)benzimidazole
Formula: C₁₀H₇N₃S
Peso molecolare: 201.25 g/mol
Punto di fusione: 305°C
MDL Number: MFCD00005587
CAS Number: 148-79-8
EINECS: 205-725-8
UN: 3077
ADR: 9 III
Merck Index: 14,09289
Visualizza i codici
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %