Stampa…

Tetramethylhydroquinone ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1,4-Dihydroxy-2,3,5,6-tetramethylbenzene, 2,3,5,6-Tetramethyl-1,4-benzenediol, 2,3,5,6-Tetramethylbenzene-1,4-diol, Durohydroquinone, Dihydroxydurene

T0822-1G T0822-5G
TCIAT0822-1GEA 214 EUR
TCIAT0822-1G TCIAT0822-5G
Tetramethylhydroquinone ≥95.0% (by GC)
Tetramethylhydroquinone
Formula: C₁₀H₁₄O₂
Peso molecolare: 166,22 g/mol
MDL Number: MFCD00045781
CAS Number: 527-18-4
Merck Index: 14,03469

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 233.0 to 237.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR