Stampa…

Trimethylolpropane phosphite ≥94.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-Ethyl-1-phospha-2,6,7-trioxabicyclo[2.2.2]octane

T0816-25G T0816-5G
TCIAT0816-25GEA 313 EUR
TCIAT0816-25G TCIAT0816-5G
Trimethylolpropane phosphite ≥94.0% (by titrimetric analysis)
Trimethylolpropane phosphite
Formula: C₆H₁₁O₃P
Peso molecolare: 162,12 g/mol
Punto di ebollizione: 100°C
Punto di fusione: 56°C
MDL Number: MFCD00152500
CAS Number: 824-11-3
EINECS: 212-523-3
UN: 3464
ADR: 6.1,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 94.0 %
Melting point 53.0 to 58.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR