Propyl-p-toluenesulphonate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
p-Toluenesulphonic acid Propyl Ester
T0726-25ML
T0726-500ML
TCIAT0726-25MLEA
29
EUR
TCIAT0726-25ML
TCIAT0726-500ML
Propyl-p-toluenesulphonate ≥98.0% (by GC)
Propyl-p-toluenesulphonate
Formula:
C₁₀H₁₄O₃S Peso molecolare: 214,28 g/mol Punto di ebollizione: 156°C |
MDL Number:
MFCD00059244 CAS Number: 599-91-7 EINECS: 209-978-5 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...