Tris(2-carboxyethyl)isocyanurate ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3,3',3''-(2,4,6-Trioxo-1,3,5-triazinane-1,3,5-triyl)tripropionic acid, Isocyanuric Acid Tris(2-carboxyethyl) Ester
T0675-25G
T0675-500G
TCIAT0675-25GEA
20
EUR
TCIAT0675-25G
TCIAT0675-500G
Tris(2-carboxyethyl)isocyanurate ≥98.0% (by titrimetric analysis)
Tris(2-carboxyethyl)isocyanurate
Formula:
C₁₂H₁₅N₃O₉ Peso molecolare: 345,26 g/mol |
MDL Number:
MFCD00059790 CAS Number: 2904-41-8 EINECS: 220-803-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 226.0 to 229.0 deg-C |
Solubility in hot Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...