Stampa…

Tetrabutylthiuram disulphide ≥98.0% (by total nitrogen basis)

Fornitore: TCI
Attenzione

Synonyms: Bis(dibutylthiocarbamoyl) Disulphide

T0635-25G T0635-500G
TCIAT0635-25GEA 16 EUR
TCIAT0635-25G TCIAT0635-500G
Tetrabutylthiuram disulphide ≥98.0% (by total nitrogen basis)
Tetrabutylthiuram disulphide
Formula: C₁₈H₃₆N₂S₄
MDL Number: MFCD00027191
CAS Number: 1634-02-2
EINECS: 216-652-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR