Triisobutylene (mixture of branched chain isomer) ≥90.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Isobutylene Trimer, Isobutene Trimer
T0456-25ML
T0456-500ML
TCIAT0456-25MLEA
16
EUR
TCIAT0456-25ML
TCIAT0456-500ML
Triisobutylene (mixture of branched chain isomer) ≥90.0% (by GC)
Triisobutylene (mixture of branched chain isomer)
Formula:
C₁₂H₂₄ Peso molecolare: 168,32 g/mol Punto di ebollizione: 177°C Flash Pt: 50°C |
MDL Number:
MFCD00060236 CAS Number: 7756-94-7 EINECS: 500-001-0 UN: 2324 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 90.0 % (total of isomers) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...