Stampa…

Tricaprin ≥98.0% (by GC)

Fornitore: TCI

Synonyms: Tridecanoin, Trisdecanoin, 1,2,3-Tridecanoylglycerol, Glycerol Tridecanoate, Propane-1,2,3-triyl tris(decanoate)

T0413-10G T0413-25G
TCIAT0413-10GEA 60 EUR
TCIAT0413-10G TCIAT0413-25G
Tricaprin ≥98.0% (by GC)
Tricaprin
Formula: C₃₃H₆₂O₆
Punto di ebollizione: 254°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00036239
CAS Number: 621-71-6
EINECS: 210-702-0

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Melting point 31.0 to 35.0 deg-C
Solubility in hot EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR