Stampa…

1,3,3-Trimethylindolino-6'-nitrobenzopyrylospiran ≥98.0% (by HPLC, titration analysis) photochromic compound

Fornitore: TCI EUROPE

Synonyms: 1',3',3'-Trimethyl-6-nitrospiro[chromene-2,2'-indoline], 1',3',3'-Trimethyl-6-nitrospiro[1(2H)-benzopyran-2,2'-indoline]

T0366-1G T0366-25G
TCIAT0366-1GEA 51 EUR
TCIAT0366-1G TCIAT0366-25G
1,3,3-Trimethylindolino-6'-nitrobenzopyrylospiran ≥98.0% (by HPLC, titration analysis) photochromic compound
1,3,3-Trimethylindolino-6'-nitrobenzopyrylospiran
Formula: C₁₉H₁₈N₂O₃
Peso molecolare: 322,36 g/mol
MDL Number: MFCD00059175
CAS Number: 1498-88-0
EINECS: 216-102-5

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR