Stampa…

m-Tolyltetrazolium red ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 2,5-Diphenyl-3-(m-tolyl)tetrazolium Chloride

T0324-1G T0324-100MG
TCIAT0324-1GEA 312 EUR
TCIAT0324-1G TCIAT0324-100MG
m-Tolyltetrazolium red ≥98.0% (by titrimetric analysis)
m-Tolyltetrazolium red
Formula: C₂₀H₁₇ClN₄
Peso molecolare: 348,83 g/mol
MDL Number: MFCD00059997
CAS Number: 88159-25-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR