Methyl-4-(chloroformyl)benzoate ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Terephthalic Acid Monomethyl Ester Chloride, 4-(Chloroformyl)benzoic Acid Methyl Ester, Methyl 4-(chlorocarbonyl)benzoate, Methyl 4-(Chlorocarbonyl)benzoate
T0283-25G
T0283-500G
TCIAT0283-25GEA
48
EUR
TCIAT0283-25G
TCIAT0283-500G
Methyl-4-(chloroformyl)benzoate ≥95.0% (by GC)
Methyl-4-(chloroformyl)benzoate
Formula:
C₉H₇ClO₃ Peso molecolare: 198,60 g/mol Punto di ebollizione: 140°C |
MDL Number:
MFCD00059486 CAS Number: 7377-26-6 EINECS: 230-937-2 UN: 3261 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 53.0 to 56.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...