Stampa…

p-Toluenesulphinic acid sodium salt ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Sodium 4-methylbenzenesulphinate, Sodium-p-toluenesulphinate, 4-Methylbenzenesulphinic acid sodium salt, Sodiumtoluene-4-sulphinate

T0275-500G T0275-25G
TCIAT0275-500GEA 211 EUR
TCIAT0275-500G TCIAT0275-25G
p-Toluenesulphinic acid sodium salt ≥98.0% (by HPLC, titration analysis)
p-Toluenesulphinic acid sodium salt
Formula: C₇H₇NaO₂S
Peso molecolare: 178,19 g/mol
MDL Number: MFCD00013136
CAS Number: 824-79-3
EINECS: 212-538-5
Merck Index: 14,09532

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Sodium hypochlorite Method) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR