Stampa…

Didodecyl-3,3'-thiodipropionate ≥90.0% (by GC)

Fornitore: TCI

Synonyms: Dilauryl 3,3'-Thiodipropionate, 3,3'-Thiodipropionic Acid Didodecyl Ester, Dilauryl thiodipropionate

T0205-25G T0205-500G
TCIAT0205-25GEA 16 EUR
TCIAT0205-25G TCIAT0205-500G
Didodecyl-3,3'-thiodipropionate ≥90.0% (by GC)
Didodecyl-3,3'-thiodipropionate
Formula: C₃₀H₅₈O₄S
MDL Number: MFCD00026589
CAS Number: 123-28-4
EINECS: 204-614-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dd5b174088ec2d14c0de2e4f5c1cfad10d77d88&vid=6dd5b174088ec2d14c0de2e4f5c1cfad10d77d88&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 90.0 %
Melting point 39.0 to 43.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR