Stampa…

2,4-Dinitrophenyl thiocyanate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2,4-Dinitrophenylrhodanide, Thiocyanic Acid 2,4-Dinitrophenyl Ester

T0199-1G T0199-25G
TCIAT0199-1GEA 24 EUR
TCIAT0199-1G TCIAT0199-25G
2,4-Dinitrophenyl thiocyanate ≥98.0% (by GC)
2,4-Dinitrophenyl thiocyanate
Formula: C₇H₃N₃O₄S
Peso molecolare: 225,18 g/mol
MDL Number: MFCD00047702
CAS Number: 1594-56-5
EINECS: 216-477-5
UN: 2811
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 138.0 to 143.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR