Stampa…

Thiamine nitrate ≥98.0% (by HPLC and UV method)

Fornitore: TCI

T0182-25G T0182-500G T0182-100G
TCIAT0182-25GEA 19 EUR
TCIAT0182-25G TCIAT0182-500G TCIAT0182-100G
Thiamine nitrate ≥98.0% (by HPLC and UV method)
Thiamine nitrate
Formula: C₁₂H₁₇N₅O₄S
MDL Number: MFCD00036330
CAS Number: 532-43-4
EINECS: 208-537-4
Merck Index: 14,09295
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cb493f3d3d678bd72b4530bf77863fc9cd200ed&vid=3cb493f3d3d678bd72b4530bf77863fc9cd200ed&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (UV-vis method) 98.0 to 102.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR