1,3,5-Triphenylformazan ≥92.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
1,3,5-Triphenyl tetrazolium formazan
T0175-1G
T0175-25G
TCIAT0175-1GEA
17
EUR
TCIAT0175-1G
TCIAT0175-25G
1,3,5-Triphenylformazan ≥92.0% (by HPLC)
1,3,5-Triphenylformazan
Formula:
C₁₉H₁₆N₄ Peso molecolare: 300,37 g/mol |
MDL Number:
MFCD00020647 CAS Number: 531-52-2 EINECS: 208-509-1 |
Risultati del test di specificazione
Appearance | Orange to Brown to Dark red powder to crystaline |
Purity (HPLC) | min. 92.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...