Stampa…

3',3'',5',5''-Tetraiodophenolphthalein ≥95.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Tetraiodophthalein, Iodophene, Iodophthalein

T0126-25G T0126-5G
TCIAT0126-25GEA 523 EUR
TCIAT0126-25G TCIAT0126-5G
3',3'',5',5''-Tetraiodophenolphthalein ≥95.0% (by HPLC, titration analysis)
3',3'',5',5''-Tetraiodophenolphthalein
Formula: C₂₀H₁₀I₄O₄
Peso molecolare: 821,91 g/mol
MDL Number: MFCD00001614
CAS Number: 386-17-4
EINECS: 206-857-9
Merck Index: 14,05038

Visualizza i codici

Risultati del test di specificazione

Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR