Tetrahydroxy-1,4-benzoquinone disodium salt
Fornitore: TCI EUROPE
Synonyms:
Sodium 4,5-Dihydroxy-3,6-dioxocyclohexa-1,4-diene-1,2-bis(olate), Tetrahydroxyquinone Disodium Salt
T0122-1G
T0122-25G
TCIAT0122-1GEA
16
EUR
TCIAT0122-1G
TCIAT0122-25G
Tetrahydroxy-1,4-benzoquinone disodium salt
Tetrahydroxy-1,4-benzoquinone disodium salt
Formula:
C₆H₄O₆ Peso molecolare: 216,06 g/mol |
MDL Number:
MFCD00036127 CAS Number: 1887-02-1 EINECS: 217-557-2 Merck Index: 14,09249 |
Risultati del test di specificazione
Appearance | Green to Dark green powder to crystal |
Absorbance | min. 0.600 (10 mg/L, HCl, 307.0 to 311.0 nm) (calcd.on dried substance) |
Solubility in dilute HCl | almost transparency |
Drying loss | max. 3.0 % |
Ignition residue (Sulfate) | 63.0 to 67.0 % (after drying) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...