Fisetin ≥96.0% (by HPLC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
3,7,3',4'-Tetrahydroxyflavone
Formula:
C₁₅H₁₀O₆ Punto di fusione: 330°C |
MDL Number:
MFCD00006829 CAS Number: 528-48-3 EINECS: 208-434-4 Merck Index: 14,04088 |
Risultati del test di specificazione
Appearance | White to Amber to Dark green powder to crystaline |
Purity (HPLC) | min. 96.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...