Stampa…

Diglycerol mixture of isomers ≥80.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Oxydi(propanediol), Tetrahydroxydipropyl Ether, 3,3'-Oxybis(propane-1,2-diol)

T0119-25G T0119-500G
TCIAT0119-25GEA 16 EUR
TCIAT0119-25G TCIAT0119-500G
Diglycerol mixture of isomers ≥80.0% (by GC)
Diglycerol
Formula: C₆H₁₄O₅
Peso molecolare: 166,17 g/mol
Flash Pt: 240°C
MDL Number: MFCD00049316
CAS Number: 627-82-7

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 80.0 % (total of isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR