Terpinyl acetate mixture of isomers ≥85.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₂H₂₀O₂ Punto di ebollizione: 94°C Flash Pt: 115°C |
MDL Number:
MFCD00037155 CAS Number: 80-26-2 EINECS: 201-265-7 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 80% (total of isomer) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...