Sildenafil citrate ≥98.0% (by HPLC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
5-[2-Ethoxy-5-[(4-methylpiperazin-1-yl)sulphonyl]phenyl]-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-7(6H)-one Citrate, 1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulphonyl]-4-methylpiperazine Citrate
Formula:
C₂₂H₃₀N₆O₄S·C₆H₈O₇ |
MDL Number:
MFCD09026931 CAS Number: 171599-83-0 Merck Index: 14,08489 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...