Solifenacin Succinate ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(S)-(R)-Quinuclidin-3-yl 1-phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate succinate
S0944-25MG
S0944-100MG
TCIAS0944-25MGEA
35
EUR
TCIAS0944-25MG
TCIAS0944-100MG
Solifenacin Succinate ≥98.0% (by HPLC)
Solifenacin Succinate
Formula:
C₂₃H₂₆N₂O₂·C₄H₆O₄ Peso molecolare: 480,56 g/mol |
MDL Number:
MFCD09952287 CAS Number: 242478-38-2 Merck Index: 14,08712 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 % (total of Ion-Pair) |
Melting point | 145.0 to 149.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...