Stampa…

Sodium Chenodeoxycholate ≥98.0% (by GC)

Fornitore: TCI
Attenzione

S0941-200MG S0941-1G
TCIAS0941-200MGEA 23 EUR
TCIAS0941-200MG TCIAS0941-1G
Sodium Chenodeoxycholate ≥98.0% (by GC)
Sodium Chenodeoxycholate
Formula: C₂₄H₃₉NaO₄
MDL Number: MFCD00065452
CAS Number: 2646-38-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5b22761284cae37b1a8a4dc3908c9346b143d30&vid=d5b22761284cae37b1a8a4dc3908c9346b143d30&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR