Stampa…

Secnidazole ≥98.0% (by GC)

Fornitore: TCI

S0911-1G S0911-5G
TCIAS0911-1GEA 18 EUR
TCIAS0911-1G TCIAS0911-5G
Secnidazole ≥98.0% (by GC)
Secnidazole
Formula: C₇H₁₁N₃O₃
MDL Number: MFCD00864656
CAS Number: 3366-95-8
EINECS: 222-134-0
Merck Index: 14,08419
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fe3b3165fbcb270de68510c4b3bb619972af93e&vid=1fe3b3165fbcb270de68510c4b3bb619972af93e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 72.0 to 76.0 deg-C
Water max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR