3'-Sialyllactose sodium salt ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
3'-Sialyllactose sodium, Sodium (2S,4S,5R,6R)-5-acetamido-2-(((2R,3S,4S,5R,6S)-3,5-dihydroxy-2-(hydroxymethyl)-6-(((2R,3R,4R,5R)-1,2,4,5-tetrahydroxy-6-oxohexan-3-yl)oxy)tetrahydro-2H-pyran-4-yl)oxy)-4-hydroxy-6-((1R,2R)-1,2,3-trihydroxypropyl)tetrahydro-2H-pyran-2-carboxylate, Neu5Ac alpha(2-3)Gal beta(1-4)Glc sodium salt
Formula:
C₂₃H₃₈NNaO₁₉ Peso molecolare: 655,53 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00166999 CAS Number: 128596-80-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) (CAD) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...