Stampa…

Sodium stearyl fumarate ≥98.0% (by titrimetric analysis)

Fornitore: TCI

S0879-25G S0879-100G
TCIAS0879-25GEA 41 EUR
TCIAS0879-25G TCIAS0879-100G
Sodium stearyl fumarate ≥98.0% (by titrimetric analysis)
Sodium stearyl fumarate
Formula: C₂₂H₄₀O₄
MDL Number: MFCD00236640
CAS Number: 4070-80-8
EINECS: 223-781-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR