N-Succinimidyl Acetate ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₆H₇NO₄ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00037916 CAS Number: 14464-29-0 EINECS: 238-454-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 132.0 to 136.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...