Stampa…

Suplatast tosylate ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: IPD 1151T, 2-[4-(3-Ethoxy-2-hydroxypropoxy)phenylcarbamoyl]ethyldimethylsulphonium p-Toluenesulphonate, Suplatast-tosylate, [3-[[4-(3-Ethoxy-2-hydroxypropoxy)phenyl]amino]-3-oxopropyl]dimethylsulphonium 4-methylbenzenesulphonate, Suplatast tosilate, (3-((4-(3-Ethoxy-2-hydroxypropoxy)phenyl)amino)-3-oxopropyl)dimethylsulphonium 4-methylbenzenesulphonate

S0875-1G S0875-100MG
TCIAS0875-1GEA 238 EUR
TCIAS0875-1G TCIAS0875-100MG
Suplatast tosylate ≥98.0% (by HPLC, total nitrogen)
Suplatast tosylate
Formula: C₂₃H₃₃NO₇S₂
Peso molecolare: 499,64 g/mol
Punto di fusione: 87°C
MDL Number: MFCD00867604
CAS Number: 94055-76-2
Merck Index: 14,09003

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR