Stampa…

Sulbactam ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Sulbactam acid, (2S,5R)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid 4,4-Dioxide

S0868-25G S0868-5G
TCIAS0868-25GEA 175 EUR
TCIAS0868-25G TCIAS0868-5G
Sulbactam ≥98.0% (by HPLC, titration analysis)
Sulbactam
Formula: C₈H₁₁NO₅S
Peso molecolare: 233,24 g/mol
Punto di fusione: 156°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00867005
CAS Number: 68373-14-8
EINECS: 269-878-2
Merck Index: 14,08889

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +233.0 to +241.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR