Stampa…

N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

Synonyms: DABCYL acid, SE, DABCYL N-Succinimidyl Ester, 4-[4-(Dimethylamino)phenylazo]benzoic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 4-((4-(dimethylamino)phenyl)diazenyl)benzoate

S0857-200MG S0857-1G
TCIAS0857-200MGEA 83 EUR
TCIAS0857-200MG TCIAS0857-1G
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥98.0% (by HPLC, titration analysis)
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate
Formula: C₁₉H₁₈N₄O₄
Peso molecolare: 366.38 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00467415
CAS Number: 146998-31-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73ba10af51ca4578fa129d59033dbb3f7a3e9e63&vid=73ba10af51ca4578fa129d59033dbb3f7a3e9e63&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR