Sodium-2,3,5,6-tetrafluoro-4-hydroxybenzenesulphonate ≥98.0% (by HPLC)
Fornitore: TCI
Synonyms:
2,3,5,6-Tetrafluoro-4-hydroxybenzenesulphonic acid Sodium Salt
Formula:
C₆H₂F₄O₄S Peso molecolare: 268.11 g/mol |
CAS Number:
221908-17-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...