Stampa…

Sodium-4-hydroxybenzenesulphonate dihydrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Phenolsulphonic acid sodium salt dihydrate, 4-Hydroxybenzenesulphonic acid sodium salt, Sodium 4-hydroxybenzenesulphonate dihydrate, Sodium p-Phenolsulphonate, p-Phenolsulphonic acid sodium salt

S0824-25G S0824-500G
TCIAS0824-25GEA 16 EUR
TCIAS0824-25G TCIAS0824-500G
Sodium-4-hydroxybenzenesulphonate dihydrate ≥98.0% (by HPLC, titration analysis)
Sodium-4-hydroxybenzenesulphonate dihydrate
Formula: C₆H₉NaO₆S
Peso molecolare: 232,19 g/mol
MDL Number: MFCD00150724
CAS Number: 10580-19-5
EINECS: 249-038-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 % (calcd.on anh.substance)
Water 14.5 to 16.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR