Stampa…

N-Succinimidyl Acrylate ≥98.0% (by GC)

Fornitore: TCI
Attenzione

S0814-25G S0814-5G
TCIAS0814-25GEA 474 EUR
TCIAS0814-25G TCIAS0814-5G
N-Succinimidyl Acrylate ≥98.0% (by GC)
N-Succinimidyl Acrylate
Formula: C₇H₇NO₄
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00078261
CAS Number: 38862-24-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR